Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein alphaisoforms XLas Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988324
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8324 Supplier Bioss Supplier No. BS8160R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mutations in GNAS gene result in pseudohypoparathyroidism type 1a, pseudohypoparathyroidism type 1b, Albright hereditary osteodystrophy, pseudopseudohypoparathyroidism, McCune-Albright syndrome, progressive osseous heteroplasia, polyostotic fibrous dysplasia of bone, and some pituitary tumors. This gene has a highly complex imprinted expression pattern. It encodes maternally, paternally, and biallelically expressed proteins which are derived from alternatively spliced transcripts with alternate 5′ exons. Each of the upstream exons is within a differentially methylated region, commonly found in imprinted genes. However, the close proximity (14 kb) of two oppositely expressed promoter regions is unusual. In addition, one of the alternate 5′ exons introduces a frameshift relative to the other transcripts, resulting in one isoform which is structurally unrelated to the others. An antisense transcript exists, and may regulate imprinting in this region. Mutations in this gene result in pseudohypoparathyroidism type 1a (PHP1a), which has an atypical autosomal dominant inheritance pattern requiring maternal transmission for full penetrance.

Specifications

Antigen G protein alphaisoforms XLas
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GNAS
Gene Accession No. O95467, P63092, P84996, Q5JWF2
Gene Alias 5530400H20Rik; A930027G11Rik; Adenylate cyclase-stimulating G alpha protein; AHO; ALEX; GNAS; NESP55; alpha-stimulatory subunit of GTP-binding protein; Alternative gene product encoded by XL-exon; BOS_13284; C130027O20Rik; C20orf45; CH242-247L10.1; dJ309F20.1.1; dJ806M20.3.3; Extra large alphas protein; G protein subunit alpha S; GAIPIRRH peptide; G-alpha-8; Galphas; Gnas; GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus; gnas {ECO:0000312; GNAS complex locus; GNAS complex locus; NESP55; GNAS1; Gnasxl; Gnpas; GPIPIRRH peptide; GPSA; Gs alpha subunit; Gsa; G-SALPHA60A; GSP; GTP-binding regulatory protein alpha subunit exon 1; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide 1; guanine nucleotide binding protein alpha s; guanine nucleotide binding protein alpha subunit; guanine nucleotide binding protein, alpha stimulating activity polypeptide 1; guanine nucleotide regulatory protein; Guanine nucleotide-binding protein G(s) subunit alpha; guanine nucleotide-binding protein G(s) subunit alpha isoforms; Guanine nucleotide-binding protein G(s) subunit alpha isoforms short; Guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas; guanine nucleotide-binding protein Gs alpha1; guanine nucleotide-binding protein G-s, alpha subunit; guanine nucleotide-binding protein, alpha-stimulating activity polypeptide 1; LHAL tetrapeptide; LSAL tetrapeptide; MGC33735; MNCb-5546; Nesp; NESP55; NESP55; guanine nucleotide-binding protein G(s) subunit alpha; Nespl; neuroendocrine secretory protein; Neuroendocrine secretory protein 55; Oedsml; Oed-Sml; OTTHUMP00000031742; OTTHUMP00000196026; OTTHUMP00000196030; P1; P2; P3; PHP1A; PHP1B; PHP1C; POH; PORGSA1; Protein ALEX; protein ALEX; protein GNAS; protein NESP55; protein ALEX; protein GNAS; protein SCG6 (secretogranin VI); RGD:2716}; RP4-543J19.4; SCG6; secretogranin VI; SgVI; stimulatory G-protein alpha subunit; stimulatory GTP binding protein; XLalphas; XLas
Gene Symbols GNAS
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human G protein alpha isoforms XLas.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2778
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.