Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-substrate Antibody (OTI2A8), Alexa Fluor™ 647, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271645AF647

Catalog No. N271645A647


Only null left
Add to Cart

Description

Description

G-substrate Monoclonal specifically detects G-substrate in Human samples. It is validated for Western Blot.
Specifications

Specifications

G-substrate
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
PPP1R17
Full length recombinant protein of human G-substrate (NP_006649) produced in E.coli.
0.1 mL
Primary
Human
IgG1
Western Blot
OTI2A8
Western Blot
chromosome 7 open reading frame 16, GSBSG-substrate
Mouse
Affinity Purified
Signal Transduction
10842.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.