Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-substrate Antibody (OTI2A8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271645
Description
G-substrate Monoclonal specifically detects G-substrate in Human samples. It is validated for Western Blot.Specifications
G-substrate | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000 | |
chromosome 7 open reading frame 16, GSBSG-substrate | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
10842 | |
Human | |
Lyophilized |
Western Blot | |
OTI2A8 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PPP1R17 | |
Full length human recombinant protein of human C7ORF16 (NP_006649) produced in E.coli. | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction