Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-substrate Antibody (OTI2A8), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271645IR
Description
G-substrate Monoclonal specifically detects G-substrate in Human samples. It is validated for Western Blot.Specifications
| G-substrate | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PPP1R17 | |
| Full length recombinant protein of human G-substrate (NP_006649) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI2A8 | |
| Western Blot | |
| chromosome 7 open reading frame 16, GSBSG-substrate | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 10842.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction