Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-substrate Antibody (OTI2A8), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271645PE
Description
G-substrate Monoclonal antibody specifically detects G-substrate in Human samples. It is validated for Western BlotSpecifications
G-substrate | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
OTI2A8 | |
Western Blot | |
chromosome 7 open reading frame 16, GSBSG-substrate | |
Full length human recombinant protein of human C7ORF16 (NP_006649) produced in E.coli. | |
0.1 mL | |
Signal Transduction | |
10842 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction