Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-substrate Antibody (OTI2E5), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271644AF532
Description
G-substrate Monoclonal specifically detects G-substrate in Human samples. It is validated for Western Blot.Specifications
G-substrate | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
PPP1R17 | |
Full length recombinant protein of human G-substrate (NP_006649) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2E5 | |
Western Blot | |
chromosome 7 open reading frame 16, GSBSG-substrate | |
Mouse | |
0.1 mL | |
10842 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction