Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ 3GX™ 19-D324 Dyneema™ Diamond Blended Gloves, Polyurethane Grip

Catalog No. 19013655 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Purple
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-013-655 X-Small Purple
19-013-656 Small Red
19-013-657 Medium Yellow
19-013-658 Large Brown
19-013-659 X-Large Black
19-013-660 2X-Large Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-013-655 Supplier PIP™ Supplier No. 19D324/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

White lightweight Dyneema™ blended gloves feature dexterity, tactile sensitivity and cut resistance. Polyurethane coated palm provides abrasion resistance, puncture resistance and secure wet or dry grip.

  • Seamless construction offers comfort and breathability
  • Lightweight shell provides dexterity, tactile sensitivity and cut resistance
  • Polyurethane coating provides a secure grip in dry and slightly wet/oily conditions and offers high dexterity and tactile sensitivity
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Color coded cuff hems for easy size identification
  • Washable, resistant to chemicals, water and ultraviolet light

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance ASTM F2992-15 ANSI Cut Level A2 (Adopted by ANSI/ISEA 02/16)/ASTM F1790-05 ANSI Cut Level 2
Certifications/Compliance (Europe) CE
Construction Seamless Knit
Coating Material Polyurethane
Color White, Black
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Dyneema, Polyurethane
Product Type Cut-resistant Gloves
Size X-Small
EN388 Standard EN 388:4542
For Use With (Application) Electronics, glass cutting, metal handling, parts assembly, construction and general duty
Glove Cuff Color Purple
Glove Cuff Style Knit
Model 19-D324/XS
Series G-Tek™ 3GX™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.