Learn More
PIP™ G-Tek™ Aramid Fiber Cut-Resistant Gloves, Nitrile Microfinish Grip
Catalog No. 19100702 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
Glove Cuff Color:
Black
Brown
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-100-702 Medium Yellow
19-100-703 Large Brown
19-100-704 X-Large Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-100-702 Supplier PIP™ Supplier No. 09K1650/M
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Cut-resistant gloves feature a nitrile microfinish coated palm and fingertips to provide an enhanced grip in dry, wet and oily conditions.
  • Longer wrist length for added protection
  • Heavier and thicker shell provides enhanced protection against cuts
  • Long-lasting and durable for great economic efficiency
  • Double dipped MicroFinish™ coating provides remarkable comfort to the wearer and prevents wet hands, keeping the inside of the glove dry
Specifications
ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Nitrile
Color Yellow, Black
Cut Resistance Rating ANSI A3
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388:4342B
For Use With (Application) Material handling, glass handling operations, fishing industries, sanitation, general maintenance, woodworking, waste handling and recycling
Glove Cuff Color Yellow
Glove Cuff Style Elasticated
Grip Design MicroFinish
Model 09-K1650/M
Puncture Rating ANSI/ISEA 105-16 Level 2
Series G-Tek™ KEV™ 09-K1650
Yarn Gauge 13