Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ ECO Series™ Seamless Knit Glove

Catalog No. 19046010 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-010 X-Large
19-046-006 X-Small
19-046-007 Small
19-046-008 Medium
19-046-009 Large
19-046-011 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-046-010 Supplier PIP™ Supplier No. 31530R/XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

G-Tek™ ECO Series™ Seamless Knit Recycled Yarn/Spandex Blended Glove with Nitrile Coated MicroSurface Grip on Palm & Fingers

  • Each pair is made from one recycled plastic bottle, reinforcing our commitment to developing innovative products that are less harmful to the environment
  • Seamless knit blended liner fibers made of 90% recycled PET water bottles and 10% Elastane for comfort, finger dexterity and breathability
  • Nitrile MicroSurface coating provides a superior grip in dry, wet and oily conditions by reacting like tiny suction cups that attach themselves firmly to the material being handled
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Breathable back for comfort, OEKO TEX approved
  • Packaged in a biodegradable poly bag of 12 pairs with technical information listed on recycled cardboard

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 388-4121
Chlorinated No
Construction Seamless Knit
Coating Material Nitrile
Color White
Exterior Finish Partially Textured
Hand Specific
Material Recycled Plastic
Product Type Seamless Knit Gloves
Size X-Large
Style Standard
For Use With (Application) Small Parts Handling, Maintenance, Inspection
Gender Unisex
Glove Cuff Style Knit
Series ECO™ Series
Yarn Gauge 13
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.