Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP™ Economy Grade Foam Nitrile Coated Nylon Gloves

Catalog No. 19149620 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-149-620 X-Small
19-149-621 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-149-620 Supplier PIP™ Supplier No. 34C232/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Seamless nylon shell increases comfort, finger dexterity, and breathability. The foam nitrile coating is designed with a cell structure to disperse fluids on contact for enhanced grip capabilities.

  • Knit wrist helps to prevent dirt and debris from entering the glove
  • Foam nitrile coating is breathable and durable
  • Gloves are resistant to chemicals, water, and ultraviolet light
  • Color coded hems for easy size identification
  • Ideal for part handling, equipment maintenance, and assembly

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile Foam
Color Gray, Black
Exterior Finish No Texture
Grade Economy
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Small
Style Standard
For Use With (Application) Manufacturing/Assembly Applications Where Abrasion, Cut and Puncture Resistance is Required
Glove Cuff Style Continuous Knit
Series 34-C232
Yarn Gauge 13
Length (English) 8.6 in.
Length (Metric) 22 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.