Learn More
PIP™ G-Tek™ Aramid Fiber Gloves, Nitrile Coated Foam Grip
Shop All Protective Industrial Products Inc. ProductsDescription
- Seamless knit construction provides comfort without sacrificing dexterity
- Gloves are inherently cut-resistant and will not melt, ignite or conduct electricity
- Foam nitrile coating is breathable and durable
- Coating is designed with a cell structure that disperses fluids on contact for improved grip
- Knit wrist helps prevent dirt and debris from entering the glove
- Launderable for extended life and reduced replacement costs
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Construction | Seamless Knit |
| Coating Material | Nitrile Foam |
| Color | Yellow, Black |
| Cut Resistance Rating | ANSI A7 |
| Hand | Specific |
| Latex Free | Yes |
| Material | Aramid Fiber, Nylon, Steel Fiber |
| Product Type | Cut-resistant Gloves |
| Size | X-Large |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.