Learn More
PIP G-Tek™ Paradox™ PolyKor™ Blended Glove with Polyurethane Coated Flat Grip on Palm and Fingers, 21 Gauge
GREENER_CHOICE
Catalog No. 01670925 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01-670-925 Small
01-670-926 X-Large
01-670-927 X-Small
01-670-928 2X-Large
01-670-929 2X-Small
01-670-931 Large
01-670-932 Medium
01-670-930 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01-670-925 Supplier PIP Supplier No. 16541/S
Add to Cart
Add to Cart
Ultra-light weight 21-gauge seamless knit gloves deliver outstanding dexterity and tactile sensitivity. Polyurethane palm coating provides a tactile grip in dry and slightly oily conditions.
  • Seamless construction offers increased comfort and breathabilityPremium PolyKor™ blended 21G shell is lightweight and provides excellent dexterity, tactile sensitivity, and cut resistance
  • Polyurethane coatings take on the surface properties of the glove liner providing a tactile grip in dry and slightly oily conditions, ideal for applications that require dexterity for precise handling
  • Touchscreen compatible to allow the user to operate a touch screen phone or device without removing gloves
  • Reinforced thumb crotch provides longer wear life and increased protection
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Washable, resistant to chemicals, water, and ultraviolet light
Specifications
Construction Seamless Knit
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material PolyKor Fiber
Product Type Cut- and Abrasion-resistant Glove
Size Small
Gender Unisex
Lining Seamless Lining
Puncture Rating ANSI/ISEA 105-16 Level 2
Thumb Style Reinforced