Learn More
PIP™ G-Tek™ PolyKor™ Gloves, Touchscreen Compatible, 18 Gauge
Catalog No. 17456181 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-456-181 X-Small
17-456-182 Small
17-456-183 Medium
17-456-184 Large
17-456-185 X-Large
17-456-186 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-456-181 Supplier PIP™ Supplier No. 16854/XS
Only null left
Add to Cart
Add to Cart
Seamless knit PolyKor™ blended glove with nitrile coated foam grip on the palm and fingers.
  • Seamless construction offers increased comfort and breathability
  • PolyKor™ shell is lightweight and provides excellent dexterity, tactile sensitivity and cut resistance
  • Nitrile is a synthetic rubber compound that offers excellent puncture, tear and abrasion resistance
  • Foam Nitrile coatings are breathable and durable, designed with a cell structure that disperse fluids on contact for an improved grip
  • Reinforced thumb crotch for extended glove life, enhanced comfort, increased abrasion resistance and added durability in a high-wear area
  • Touchscreen compatible to allow the user to operate a touch screen phone or device without removing gloves
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Color coded hems for easy size identification
  • Washable, resistant to chemicals, water, and ultraviolet light
Specifications
Construction Seamless Knit
Coating Material Nitrile Foam
Color Salt and Pepper, Black
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free No
Material PolyKor Fiber
Product Type Cut-resistant Gloves
Size X-Small
EN388 Standard EN388:4X42D
Gender Unisex
Glove Cuff Color Purple
Glove Cuff Style Knitwrist
Lining Seamless Lining
Puncture Rating ANSI/ISEA 105-16 Level 3
Yarn Gauge 18