Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ VR-X™ Cut Resistant Seamless Knit Glove

Catalog No. 19270041 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-270-041 Small
19-270-040 X-Small
19-270-042 Medium
19-270-043 Large
19-270-044 X-Large
19-270-045 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-270-041 Supplier PIP™ Supplier No. 16VRX380/S
Add to Cart
Add to Cart
This item is not returnable. View return policy

G-Tek™ VR-X™ Cut Resistant Seamless Knit PolyKor™ Blended Glove with Polyurethane Advanced Barrier Coating Protection with Touchscreen Compatibility

  • Seamless construction offers increased comfort and breathability
  • VR-X Polyurethane coatings provide advanced barrier protection, the treated coating takes on the surface properties of the glove liner providing a tactile grip in wet, dry and slightly oily conditions, ideal for applications that require dexterity for precise handling
  • Liquid proof, air leak tested - EN374-2
  • Antibacterial and antiviral barrier protection per ISO 20743:2013 test method & EN374-5
  • 4.0 AQL, similar to the AQL levels for industrial grade disposable gloves
  • ESD Safe, Surface Resistance per ESD S11.11 Static dissipative range at 109 ohms, 12% RH, 48 hr. at 100 Volts
  • Touchscreen compatible to allow the user to operate a touch screen phone or device without removing gloves
  • 100% Silicone free
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Color coded hems for easy size identification
  • Washable, resistant to water and ultraviolet light
  • Recommended wash cycle: maximum 6 cycles

Specifications

ANSI Abrasion Rating ANSI 1
Certifications/Compliance ANSI Abrasion 1, ANSI Cut A3
Certifications/Compliance (Europe) EN 388- 4X32C, EN374-2, EN374-5
Chlorinated No
Construction Seamless Knit
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A3
Exterior Finish Partially Textured
Grade Industrial
Hand Specific
Material HPPE
Product Type Seamless Knit Gloves
Size Small
Style Standard
For Use With (Application) Small Parts Handling, Maintenance and Repair Operations
Gender Unisex
Glove Cuff Style Knit
Yarn Gauge 18
Length (English) 10 in.
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.