Learn More
CERILLIANT™ Gabapentin-D10 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco G0171ML
Description
Sold under the trade names Neurontin™, Nupentin, or Gabarone, gabapentin is an anticonvulsant drug used primarily for treatment of seizures, fibromyalgia, and chronic neuropathic pain. This internal standard is suitable for GC/MS or LC/MS testing or isotope dilution methods of gabapentin in forensic analysis, clinical toxicology, pain prescription monitoring, or urine drug testing.
Specifications
| 67-56-1 , 1126623-20-8 | |
| MFCD16294730 | |
| 1.0 mg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
| C9D10H7NO2 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 181.30 | |
| 1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
CERILLIANT is a registered trademark of Cerilliant Corporation