Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G0S2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306188JF646
Description
G0S2 Polyclonal specifically detects G0S2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
G0S2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
G0/G1 switch regulatory protein 2, G0/G1switch 2, RP1-28O10.2 | |
Produced in rabbits immunized with E. coli-derived Human G0S2 fragment. | |
0.1 mL | |
Cell Cycle and Replication | |
50486 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction