Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ G0S2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523087
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523087 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523087 Supplier Invitrogen™ Supplier No. PA523087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G0S2 Polyclonal Antibody for ICC/IF

The G(0)-G(1) switch gene 2 (G0S2) codes for a mitochondrial protein that is important in apoptosis, lipolysis, and triacylglycerol hydrolysis. In adipocytes, G0S2 binds to adipose triglyceride lipase and decreases the breakdown of lipid droplets. In Crohn's colitis, the presence of G0S2 is increased in correlation to the amount of non-converted lipid in waste. In squamous lung cancer, mutations in the G0S2 gene are common. In breast cancer, epigenetic silencing of G0S2 occurs, and normal apoptotic regulation of aberrant cells is negatively impacted. However, when G0S2 is not epigenetically modified, it is the best mediator of the Bcl-2/Bax heterodimer, resulting in apoptosis of cancerous cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G0S2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 50% glycerol and 0.05% sodium azide
Gene G0s2
Gene Accession No. P27469
Gene Alias AI255151; AV006465; G0/G1 switch 2; G0/G1 switch gene 2; G0/G1 switch protein 2; G0/G1 switch regulatory protein 2; G0/G1switch 2; G0s2; G0S2-like protein; putative lymphocyte G0/G1 switch; Putative lymphocyte G0/G1 switch gene; putative lymphocyte G0/G1 switch protein 2; RP1-28O10.2
Gene Symbols G0s2
Host Species Rabbit
Immunogen A genomic peptide made to the human G0S2 protein (residues 1-103).
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50486
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.