Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G0S2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | G0S2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
G0S2 Polyclonal specifically detects G0S2 in Human samples. It is validated for Western Blot.Specifications
G0S2 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
G0/G1 switch regulatory protein 2, G0/G1switch 2, RP1-28O10.2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human G0S2 (NP_056529.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS with 50% glycerol, pH7.3. | |
50486 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title