Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G2 and S phase expressed 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987868
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7868 Supplier Bioss Supplier No. BS7611R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is only expressed in the S and G2 phases of the cell cycle, where it colocalizes with cytoplasmic tubulin and microtubules. In response to DNA damage, the encoded protein accumulates in the nucleus and binds the tumor suppressor protein p53, shuttling it out of the nucleus and repressing its ability to induce apoptosis.

Specifications

Antigen G2 and S phase expressed 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GTSE1
Gene Accession No. Q9NYZ3
Gene Alias B99; G2 and S phase-expressed protein 1; G-2 and S-phase expressed 1; G2 and S-phase expressed 1; GTSE1; GTSE-1; Protein B99 homolog
Gene Symbols GTSE1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GTSE1/G2 and S phase expressed 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51512
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.