Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ G2E3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569221
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Essential in early embryonic development to prevent apoptotic death.
Specifications
G2E3 | |
Polyclonal | |
Unconjugated | |
G2E3 | |
6030408C04Rik; 9030416F18; AW046379; D930034K21Rik; G2/M phase-specific E3 ubiquitin-protein ligase; G2/M phase-specific HECT-type E3 ubiquitin transferase; G2/M-phase specific E3 ubiquitin ligase; G2/M-phase specific E3 ubiquitin protein ligase; G2e3; hypothetical protein LOC508575; KIAA1333; mKIAA1333; PHD finger protein 7B; PHF7B; RGD1310263 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55632 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7L622 | |
G2E3 | |
synthetic peptide directed towards the N terminal region of human G2E3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction