Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ G2E3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589808 Supplier Invitrogen™ Supplier No. PA589808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNESKPGDSQ NLACVFCRKH DDCPNKYGEK KTKEKWNLTV HYYCLLMSSG IWQRGKEEEG VYGFLIEDIR KEVNRASKLK CCVCKKNGAS IGCVAPRCKR SYHFPCGLQR ECIFQFTGNF ASFCWDHRPV QIITSNNYRE SLPCTICLEF IEPIPSYNIL RSPCCKNAWF HRDCLQVQAI; Positive Samples: U-87MG, LO2, 293T, OVCAR3, Mouse eye, Mouse brain, Mouse liver, Rat testis; Cellular Location: Cytoplasm, Nucleus, nucleolus.

E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Essential in early embryonic development to prevent apoptotic death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G2E3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene G2E3
Gene Accession No. Q5RJY2, Q7L622
Gene Alias 6030408C04Rik; 9030416F18; AW046379; D930034K21Rik; G2/M phase-specific E3 ubiquitin-protein ligase; G2/M phase-specific HECT-type E3 ubiquitin transferase; G2/M-phase specific E3 ubiquitin ligase; G2/M-phase specific E3 ubiquitin protein ligase; G2e3; hypothetical protein LOC508575; KIAA1333; mKIAA1333; PHD finger protein 7B; PHF7B; RGD1310263
Gene Symbols G2E3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human G2E3 (NP_0602392).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217558, 299002, 55632
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.