Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ G2E3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551610 Supplier Invitrogen™ Supplier No. PA551610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IQAYPEAFCS ILCHKPESLS AKILSELFTV HTLPDVKALG FWNSYLQAVE DGKSTTTMED ILIFATGCSS IPPAGFKPTP SIECLHVDFP VGNKCNNCLA IPITNTYKEF QENMDFTIRN T Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 81%.

E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Essential in early embryonic development to prevent apoptotic death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G2E3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene G2E3
Gene Accession No. Q7L622
Gene Alias 6030408C04Rik; 9030416F18; AW046379; D930034K21Rik; G2/M phase-specific E3 ubiquitin-protein ligase; G2/M phase-specific HECT-type E3 ubiquitin transferase; G2/M-phase specific E3 ubiquitin ligase; G2/M-phase specific E3 ubiquitin protein ligase; G2e3; hypothetical protein LOC508575; KIAA1333; mKIAA1333; PHD finger protein 7B; PHF7B; RGD1310263
Gene Symbols G2E3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human G2E3. Recombinant protein control fragment (Product #RP-88865).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55632
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.