Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G2E3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | G2E3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
G2E3 Polyclonal specifically detects G2E3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
G2E3 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
55632 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EC 6.3.2, EC 6.3.2.-, FLJ20333, G2/M phase-specific E3 ubiquitin-protein ligase, G2/M-phase specific E3 ubiquitin ligase, G2/M-phase specific E3 ubiquitin protein ligase, KIAA1333PHD finger protein 7B, PHF7B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human G2E3 (NP_060239.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title