Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G3BP1, Mouse anti-Human, Clone: 2F3, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation H00010146M01J
Description
Sequence: KPEPVLEETAPEDAQKSSSPAPADIAQTVQEDLRTFSWASVTSKNLPPSGAVPVTGIPPHVVKVPASQPRPESKPESQIPPQRPQRDQRVSpecifications
G3BP1 | |
Monoclonal | |
Unconjugated | |
GTPase activating protein (SH3 domain) binding protein 1 | |
G3BP/HDH-VIII/MGC111040 | |
Mouse | |
Affinity chromatography | |
RUO | |
10146 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2F3 | |
PBS with no preservative; pH 7.4 | |
BC006997 | |
G3BP1 | |
G3BP1 (AAH06997, 214 a.a. ∼ 303 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction