Learn More
Invitrogen™ G3BP1 Polyclonal Antibody

Description
Immunogen sequence: FRYQDEVFGG FVTEPQEESE EEVEEPEERQ QTPEVVPDDS GTFYDQAVVS NDMEEHLEEP VAEPEPDPEP EPEQEPVSEI QEEKPEPVLE ETAP Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 87%.
Specifications
Specifications
| Antigen | G3BP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | G3BP1 |
| Gene Accession No. | P97855, Q13283 |
| Gene Alias | AI849976; ATP-dependent DNA helicase VIII; B430204O07; C87777; DNA helicase VIII; G3BP; G3BP stress granule assembly factor 1; G3bp1; G3BP-1; GAP binding protein; GAP SH3 binding protein; GAP SH3 domain-binding protein 1; GTPase activating protein (SH3 domain) binding protein 1; hDH VIII; HDH-VIII; MGC111040; mKIAA4115; Ras GTPase-activating protein-binding protein 1; RasGAP-associated endoribonuclease G3BP; Ras-GTPase-activating protein SH3-domain binding protein 1; Ras-GTPase-activating protein SH3-domain-binding protein; RGD1310666 |
| Gene Symbols | G3BP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.