Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G3BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113642
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113642 Supplier Thermo Scientific Supplier No. PA5113642
Add to Cart
Add to Cart

G3BP1 Polyclonal Antibody for Western Blot, IHC (P), IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen G3BP1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene G3BP1
Gene Accession No. Q13283
Gene Alias AI849976; ATP-dependent DNA helicase VIII; B430204O07; C87777; DNA helicase VIII; G3BP; G3BP stress granule assembly factor 1; G3bp1; G3BP-1; GAP binding protein; GAP SH3 binding protein; GAP SH3 domain-binding protein 1; GTPase activating protein (SH3 domain) binding protein 1; hDH VIII; HDH-VIII; MGC111040; mKIAA4115; Ras GTPase-activating protein-binding protein 1; RasGAP-associated endoribonuclease G3BP; Ras-GTPase-activating protein SH3-domain binding protein 1; Ras-GTPase-activating protein SH3-domain-binding protein; RGD1310666
Gene Symbols G3BP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human G3BP1 (aa 107-156).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10146, 171092
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.