Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ G3BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553776
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553776 Supplier Invitrogen™ Supplier No. PA553776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HNDMFRYEDE VFGDSEPELD EESEDEVEEE QEERQPSPEP VQENANSGYY EAHPVTNGIE EPLEESSHEP EPEPESETKT EELKPQVE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

G3BP2 gene ontology annotations related to this gene include stress granule assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G3BP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene G3BP2
Gene Accession No. Q9UN86
Gene Alias AA409541; E430034L04Rik; G3BP; G3BP stress granule assembly factor 2; G3bp2; G3BP-2; GAP SH3 domain-binding protein 2; GTPase activating protein (SH3 domain) binding protein 2; KIAA0660; mKIAA0660; Ras GTPase-activating protein-binding protein 2; Ras-GTPase activating protein SH3 domain-binding protein 2; RGD1309571
Gene Symbols G3BP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human G3BP2. Recombinant protein control fragment (Product #RP-91849).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9908
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.