Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G418 Disulfate, Cell Culture Reagent, PROMO, Thermo Scientific Chemicals

Catalog No. J6267114PM
Click to view available options
Quantity:
25 g

G418 sulfate, geneticin is an antibiotic closely related to Gentamicin B, the common aminoglycoside antibiotic.

General description

  • G418 sulfate (geneticin) is an aminoglycoside antibiotic isolated from Micromonospora rhodorangea
  • G418 is a bacteriostatic that prevents polypeptide formation by inhibiting the elongation step of polypeptide synthesis

    Applications

  • G418 sulfate can be used for selection of genetically engineered cells in both eukaryotes and prokaryotes
  • Laboratory grade for applications such as biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20H44N4O18S2
Molecular Weight (g/mol) 692.7 g/mol
MDL Number MFCD00058314
InChI Key UHEPSJJJMTWUCP-KETIWVBHNA-N
Synonym Geneticin disulfate
PubChem CID 134129582
IUPAC Name (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
SMILES OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O

Specifications

Type G418 Disulfate
Physical Form Powder
Color White
Quantity 25 g
Solubility Information Soluble in water.
Grade Cell Culture
Chemical Name or Material G418 disulfate

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.