Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G418 Sulfate (White to Off-white Powder), Fisher BioReagents

Catalog No. p-26034 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20H44N4O18S2
Molecular Weight (g/mol) 692.70
MDL Number MFCD00058314
InChI Key UHEPSJJJMTWUCP-KETIWVBHNA-N
PubChem CID 134129582
IUPAC Name (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
SMILES OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O

Specifications

Type G418 Sulfate
Physical Form Solid
Absorbance < 0.015 at 280nm, < 0.10 at 570nm
Packaging Amber Glass
Potency > 730μg/mg
Water 10% Max.
Color White
Quantity 1 g
Assay Percent Range > 98 %
Formula Weight 692.7
Percent Purity >98%
Chemical Name or Material G418 Sulfate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
May impair fertility. May cause skin, eye, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe vapors/dust. Do not ingest. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.