Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ G6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587682 Supplier Invitrogen™ Supplier No. PA587682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NPGASLDGRP GDRVNLSCGG VSHPIRWVWA PSFPACKGLS KGRRPILWAS SSGTPTVPPL QPFVGRLRSL DSGIRRLELL LSAGDSGTFF CKGRHEDESR TVLHVLGDRT YCKAPGPTHG SVYPQ; Positive Samples: K-562, Jurkat, SW620, Mouse intestine, Mouse pancreas, Rat pancreas; Cellular Location: Cell membrane, Endoplasmic reticulum, Golgi apparatus, Golgi apparatus, Single-pass type I membrane protein.

G6B is a member of the immunoglobulin (Ig) superfamily with 241 amino acids and is a glycosylated, plasma membrane-bound cell surface receptor, but soluble isoforms encoded by some transcript variants have been found in the endoplasmic reticulum and Golgi before being secreted. Multiple transcript variants encoding different isoforms have been reported. RT-PCR, Western blot, and flow cytometric analyses suggest that G6B is usually expressed on platelets and cross linking of G6B inhibits platelet aggregation and activation in a calcium-independent manner by agonists such as ADP and collagen-related peptide. G6B is an inhibitory surface receptor on platelets and may be an antithrombotic drug target. Isoform B is a putative inhibitory receptor. Isoform A may be its activating counterpart. It is known to bind to heparin. G6B is usually expressed in a restricted set of hematopoietic cell lines like K562, Molt4 and Jurkat and not detected in the cell lines like U937, Raji, Tk, HeLa, NKL, NK62 and YT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G6B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mpig6b
Gene Accession No. D7PDD4, O95866, Q6MG59
Gene Alias AU023871; C6orf25; chromosome 6 open reading frame 25; DADB-110M10.2; G6b; G6B-B; immunoglobulin receptor; immunoreceptor tyrosine-based inhibitory motif (ITIM) containing platelet receptor; Immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing platelet receptor; ITIM-receptor G6b-B; Megakaryocyte and platelet inhibitory receptor G6b; MPIG6B; NG31; Protein G6b
Gene Symbols Mpig6b
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106722, 406865, 80739
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.