Learn More
Invitrogen™ G6B Polyclonal Antibody

Description
G6B Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | G6B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | Mpig6b |
| Gene Accession No. | O95866 |
| Gene Alias | AU023871; C6orf25; chromosome 6 open reading frame 25; DADB-110M10.2; G6b; G6B-B; immunoglobulin receptor; immunoreceptor tyrosine-based inhibitory motif (ITIM) containing platelet receptor; Immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing platelet receptor; ITIM-receptor G6b-B; Megakaryocyte and platelet inhibitory receptor G6b; MPIG6B; NG31; Protein G6b |
| Gene Symbols | Mpig6b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.