Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G6PC Antibody (CL5818), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | G6PC |
---|---|
Clone | CL5818 |
Dilution | Immunohistochemistry-Paraffin 1:5000 - 1:10000 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
G6PC Monoclonal specifically detects G6PC in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
G6PC | |
Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
Unconjugated | |
Mouse | |
EC 3.1.3.9, G6Pase, G-6-Pase, G6Pase-alpha, G6PT, glucose-6-phosphatase, Glucose-6-phosphatase alpha, glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierkedisease), glucose-6-phosphatase, catalytic subunit, GSD1, GSD1a, MGC163350 | |
G6PC | |
IgG2b | |
Protein A purified |
CL5818 | |
Monoclonal | |
Purified | |
Human | |
2538 | |
This antibody was developed against a synthetic peptide corresponding to amino acids: DFGIQSTHYLQVNYQDSQD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title