Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ G6PC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100249
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100249 Supplier Invitrogen™ Supplier No. PA5100249
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total G6PC.

Glucose-6-phosphatase is an integral membrane protein of the endoplasmic reticulum that catalyzes the hydrolysis of D-glucose 6-phosphate to D-glucose and orthophosphate. It is a key enzyme in glucose homeostasis, functioning in gluconeogenesis and glycogenolysis. Defects in the enzyme cause glycogen storage disease type I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G6PC
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene G6PC
Gene Accession No. P35575
Gene Alias AW107337; G6Pase; G-6-Pase; G6Pase-alpha; G6PC; G6PC1; G6PT; Glc-6-Pase; Glc-6-Pase-alpha; glucose-6-phosphatase; Glucose-6-phosphatase alpha; glucose-6-phosphatase catalytic subunit; glucose-6-phosphatase, catalytic; glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierke disease); glucose-6-phosphatase, catalytic subunit; GSD1; GSD1a; MGC163350; Unknown (protein for MGC:137220)
Gene Symbols G6PC
Host Species Rabbit
Immunogen A synthesized peptide derived from human G6PC(Accession P35575), corresponding to amino acid residues G68-G118.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2538
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.