Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G6PC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570652 Supplier Thermo Scientific Supplier No. PA570652
Add to Cart
Add to Cart

G6PC3 Polyclonal Antibody

G6PC3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen G6PC3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene G6PC3
Gene Accession No. Q6NSQ9
Gene Alias 0710001K01Rik; 3.1.3.9; AU019276; AU045429; AV128920; AW545836; G6Pase 3; G-6-Pase 3; G6Pase-beta; G6pc3; glucose 6 phosphatase catalytic subunit 3; glucose 6 phosphatase, catalytic, 3; glucose-6-phosphatase 3; Glucose-6-phosphatase beta; glucose-6-phosphatase catalytic subunit 3; glucose-6-phosphatase catalytic subunit-related; glucose-6-phosphatase catalytic subunit-related protein; SCN4; Ubiquitous glucose-6-phosphatase catalytic subunit-related protein; ubiquitously expressed G6Pase catalytic subunit-related protein; UGRP; zgc:158425
Gene Symbols G6PC3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse G6PC3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68401
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.