Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ G6PC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109749 Supplier Invitrogen™ Supplier No. PA5109749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G6PC3 Polyclonal Antibody for ELISA, ICC/IF, Western Blot

Hydrolyzes glucose-6-phosphate to glucose in the endoplasmic reticulum. May form with the glucose-6-phosphate transporter (SLC37A4/G6PT) a ubiquitously expressed complex responsible for glucose production through glycogenolysis and gluconeogenesis. Probably required for normal neutrophil function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G6PC3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene G6PC3
Gene Accession No. Q6AZ83, Q6NSQ9, Q9BUM1
Gene Alias 0710001K01Rik; 3.1.3.9; AU019276; AU045429; AV128920; AW545836; G6Pase 3; G-6-Pase 3; G6Pase-beta; G6pc3; glucose 6 phosphatase catalytic subunit 3; glucose 6 phosphatase, catalytic, 3; glucose-6-phosphatase 3; Glucose-6-phosphatase beta; glucose-6-phosphatase catalytic subunit 3; glucose-6-phosphatase catalytic subunit-related; glucose-6-phosphatase catalytic subunit-related protein; SCN4; Ubiquitous glucose-6-phosphatase catalytic subunit-related protein; ubiquitously expressed G6Pase catalytic subunit-related protein; UGRP; zgc:158425
Gene Symbols G6PC3
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303565, 68401, 92579
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.