Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ G6PD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581862 Supplier Invitrogen™ Supplier No. PA581862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVRLQFHDVA GDIFHQQCKR NELVIRVQPN EAVYTKMMTK KPGMFFNPEE SELDLTYGNR YKNVKLPDAY ERLILDVFCG SQMHFVRSDE LREAWRIFTP LLHQIELEKP Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene encodes glucose-6-phosphate dehydrogenase. This protein is a cytosolic enzyme encoded by a housekeeping X-linked gene whose main function is to produce NADPH, a key electron donor in the defense against oxidizing agents and in reductive biosynthetic reactions. G6PD is remarkable for its genetic diversity. Many variants of G6PD, mostly produced from missense mutations, have been described with wide ranging levels of enzyme activity and associated clinical symptoms. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G6PD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene G6PD
Gene Accession No. P11413
Gene Alias G28A; G6PD; G6PD1; G6pd-1; G6pdx; Glucose-6-phosphate 1-dehydrogenase; glucose-6-phosphate 1-dehydrogenase X; glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase X-linked; Gpdx
Gene Symbols G6PD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human G6PD. Recombinant protein control fragment (Product #RP-91906).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2539
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.