Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ G6PD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590417 Supplier Invitrogen™ Supplier No. PA590417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QIYRIDHYLG KEMVQNLMVL RFANRIFGPI WNRDNIACVI LTFKEPFGTE GRGGYFDEFG IIRDVMQNHL LQMLCLVAME KPASTNSDDV RDEKVKVLKC ISEVQANNVV LGQYVGNPDG EGEATKGYLD DPTVPRGSTT ATFAAVVLYV ENERWDGVPF ILRCGKALNE RKAEVRLQFH DVAGDIFHQQ CKRNELVIRV QPNEAVYTKM MTKKPGMFFN PEESELDLTY GNRYKNVKLP DAYERLILDV FCGSQMHFVR SDELREAWRI FTPLLHQIEL EKPKPIPYIY GSRGPTEADE LMKRVGFQYE GTYKWVNPHK L Positive Samples: MCF7, A-549, NCI-H460, HT-29, K-562, HeLa, 293T, Mouse spleen.

This gene encodes glucose-6-phosphate dehydrogenase. This protein is a cytosolic enzyme encoded by a housekeeping X-linked gene whose main function is to produce NADPH, a key electron donor in the defense against oxidizing agents and in reductive biosynthetic reactions. G6PD is remarkable for its genetic diversity. Many variants of G6PD, mostly produced from missense mutations, have been described with wide ranging levels of enzyme activity and associated clinical symptoms. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G6PD
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene G6PD
Gene Accession No. P05370, P11413, Q00612
Gene Alias G28A; G6PD; G6PD1; G6pd-1; G6pdx; Glucose-6-phosphate 1-dehydrogenase; glucose-6-phosphate 1-dehydrogenase X; glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase X-linked; Gpdx
Gene Symbols G6PD, G6pdx
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14381, 24377, 2539
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.