Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ G6PD Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $482.00
Specifications
Antigen | G6PD |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119138
|
Invitrogen™
PA5119138 |
200 μL |
Each for $482.00
|
|
|||||
PIPA5121973
|
Invitrogen™
PA5121973 |
20 μL |
Each for $178.40
|
|
|||||
Description
G6PD Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes glucose-6-phosphate dehydrogenase. This protein is a cytosolic enzyme encoded by a housekeeping X-linked gene whose main function is to produce NADPH, a key electron donor in the defense against oxidizing agents and in reductive biosynthetic reactions. G6PD is remarkable for its genetic diversity. Many variants of G6PD, mostly produced from missense mutations, have been described with wide ranging levels of enzyme activity and associated clinical symptoms. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia. Two transcript variants encoding different isoforms have been found for this gene.Specifications
G6PD | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
G6PD | |
G28A; G6PD; G6PD1; G6pd-1; G6pdx; Glucose-6-phosphate 1-dehydrogenase; glucose-6-phosphate 1-dehydrogenase X; glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase X-linked; Gpdx | |
G6PD | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P11413 | |
2539 | |
Recombinant protein Glucose-6-phosphate Dehydrogenase. The antigen corresponds to amino acid range 1-515 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title