Learn More
Blackline Safety™ G7x Satellite-Connected Safety Monitor with Multi-Gas Pump Sensor Cartridge, North America
Catalog No. 170000074 Shop All Blackline Safety Corporation Products
Change view
Click to view available options
Components Measured:
Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, PID
Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P
Hydrogen Sulfide, LEL-I
Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-000-0074 Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, PID
17-000-0143 Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen
17-000-0144 Hydrogen Sulfide, LEL-I
17-000-0146 Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-000-0074 Supplier Blackline Safety™ Supplier No. G7XPDOPVNA
Wireless connectivity ensures that each G7x is remotely configurable and firmware updates are automatic. G7x supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
  • Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
  • Emergency latch: Pull latch to trigger remote alert
  • Silent emergency: Press and hold latch to trigger remote alert
  • Low-battery: configurable threshold
  • Worker check-in: Configurable timer (5 to 180 min, or off ), automatic check-in when driving
Specifications
Description G7x 900MHz
Components Measured Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, PID
Type Multigas Cartridge