Learn More
Blackline Safety™ G7c Cellular-Connected Safety Monitor with Single-Gas Sensor Cartridge, North America
Shop All Blackline Safety Corporation ProductsDescription
- Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
- Emergency latch: Pull latch to trigger remote alert
- Silent emergency: Press and hold latch to trigger remote alert
- Low-battery: configurable threshold
- Worker check-in: Configurable timer (5 to180 min, or off ), automatic check-in when driving
Specifications
Specifications
| Components Measured | Carbon Dioxide |
| Type | Single Gas Cartridge |