Learn More
Blackline Safety™ G7c Cellular-Connected Safety Monitor with Single-Gas Sensor Cartridge, North America
Catalog No. 170000065 Shop All Blackline Safety Corporation Products
Change view
Click to view available options
Components Measured:
Ammonia
Carbon Dioxide
Carbon Monoxide
Carbon Monoxide - H
Chlorine
Hydrogen Cyanide
Hydrogen Sulfide
Oxygen
PID
Sulfur Dioxide
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-000-0065 Oxygen
17-000-0060 Ammonia
17-000-0061 Carbon Dioxide
17-000-0062 Hydrogen Sulfide
17-000-0063 Chlorine
17-000-0064 Carbon Monoxide
17-000-0066 Carbon Monoxide - H
17-000-0067 Sulfur Dioxide
17-000-0068 PID
17-000-0072 Hydrogen Cyanide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17-000-0065 Supplier Blackline Safety™ Supplier No. G7CSONA
Wireless connectivity ensures that each G7c is remotely configurable and firmware updates are automatic. G7c supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
  • Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
  • Emergency latch: Pull latch to trigger remote alert
  • Silent emergency: Press and hold latch to trigger remote alert
  • Low-battery: configurable threshold
  • Worker check-in: Configurable timer (5 to180 min, or off ), automatic check-in when driving
Specifications
Components Measured Oxygen
Type Single Gas Cartridge