Learn More
- Home
- Products
- Industrial Hygiene and Environmental Monitoring
- Gas and Vapor Detection
- Multigas Portable Detectors
- Blackline Safety™ G7c Cellular-Connected Safety Monitor with Multi-Gas Sensor Cartridge, North America
Blackline Safety™ G7c Cellular-Connected Safety Monitor with Multi-Gas Sensor Cartridge, North America
Catalog No. 170000002 Shop All Blackline Safety Corporation Products
Change view
Click to view available options
Components Measured:
Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen
Carbon Dioxide, LEL-I, Carbon Monoxide, Oxygen
Carbon Dioxide, LEL-I, Oxygen
Carbon Dioxide, Oxygen
Carbon Dioxide, Oxygen, LEL-P
Carbon Monoxide, Oxygen
Carbon Monoxide, Oxygen, LEL-P
Carbon Monoxide, Oxygen, LEL-P, PID
Chlorine, Carbon Monoxide, Oxygen, LEL-P
Chlorine, Oxygen, LEL-P, Cobalt Hydride
Hydrogen Sulfide, Carbon Monoxide
Hydrogen Sulfide, Carbon Monoxide, Oxygen
Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P
Hydrogen Sulfide, LEL-I, Chlorine, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen, Carbon Monoxide - H
Hydrogen Sulfide, LEL-I, Oxygen, PID
Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide
Hydrogen Sulfide, Oxygen, Carbon Monoxide - H
Hydrogen Sulfide, Oxygen, LEL-P
Hydrogen Sulfide, Oxygen, LEL-P, Carbon Monoxide - H
LEL-I, Carbon Monoxide, Oxygen
LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide
LEL-I, Chlorine, Carbon Monoxide, Oxygen
LEL-I, Chlorine, Oxygen
LEL-I, Chlorine, Oxygen, Sulfur Dioxide
LEL-I, Oxygen
LEL-I, Oxygen, PID
LEL-I, PID
Methane, Carbon Dioxide
Methane, Carbon Dioxide, LEL-I, Oxygen
Methane, Carbon Dioxide, Oxygen
Methane, Carbon Dioxide, Oxygen, LEL-P
Methane, Carbon Monoxide
Methane, Carbon Monoxide, Oxygen, LEL-P
Methane, Hydrogen Sulfide, LEL-I, Oxygen
Methane, Hydrogen Sulfide, Oxygen
Methane, LEL-I, Carbon Monoxide, Oxygen
Methane, LEL-I, Chlorine, Oxygen
Methane, LEL-I, Oxygen
Methane, LEL-I, Oxygen, Cobalt Hydride
Methane, LEL-I, Oxygen, Sulfur Dioxide
Methane, Oxygen
Oxygen, LEL-P
Oxygen, LEL-P, Cobalt Hydride, PID
Oxygen, LEL-P, PID
Oxygen, PID
48 product options available for selection
| Catalog No. | Components Measured |
|---|---|
| 17-000-0002 | Methane, Carbon Dioxide, LEL-I, Oxygen |
| 17-000-0003 | Methane, Carbon Dioxide, Oxygen, LEL-P |
| 17-000-0004 | Methane, Carbon Dioxide, Oxygen |
| 17-000-0005 | Methane, Carbon Dioxide |
| 17-000-0008 | Methane, Hydrogen Sulfide, LEL-I, Oxygen |
| 17-000-0009 | Methane, Hydrogen Sulfide, Oxygen |
| 17-000-0010 | Methane, LEL-I, Chlorine, Oxygen |
| 17-000-0011 | Methane, LEL-I, Carbon Monoxide, Oxygen |
| 17-000-0012 | Methane, LEL-I, Oxygen, Cobalt Hydride |
| 17-000-0013 | Methane, LEL-I, Oxygen, Sulfur Dioxide |
| 17-000-0014 | Methane, LEL-I, Oxygen |
| 17-000-0015 | Methane, Carbon Monoxide, Oxygen, LEL-P |
| 17-000-0016 | Methane, Carbon Monoxide |
| 17-000-0017 | Methane, Oxygen |
| 17-000-0020 | Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen |
| 17-000-0021 | Carbon Dioxide, LEL-I, Carbon Monoxide, Oxygen |
| 17-000-0022 | Carbon Dioxide, LEL-I, Oxygen |
| 17-000-0024 | Carbon Dioxide, Oxygen, LEL-P |
| 17-000-0025 | Carbon Dioxide, Oxygen |
| 17-000-0031 | Hydrogen Sulfide, LEL-I, Chlorine, Oxygen |
| 17-000-0033 | Hydrogen Sulfide, LEL-I, Oxygen, Carbon Monoxide - H |
| 17-000-0034 | Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide |
| 17-000-0035 | Hydrogen Sulfide, LEL-I, Oxygen, PID |
| 17-000-0036 | Hydrogen Sulfide, LEL-I, Oxygen |
| 17-000-0037 | Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P |
| 17-000-0038 | Hydrogen Sulfide, Carbon Monoxide, Oxygen |
| 17-000-0039 | Hydrogen Sulfide, Carbon Monoxide |
| 17-000-0040 | Hydrogen Sulfide, Oxygen, LEL-P, Carbon Monoxide - H |
| 17-000-0041 | Hydrogen Sulfide, Oxygen, LEL-P |
| 17-000-0042 | Hydrogen Sulfide, Oxygen, Carbon Monoxide - H |
| 17-000-0043 | LEL-I, Chlorine, Carbon Monoxide, Oxygen |
| 17-000-0044 | LEL-I, Chlorine, Oxygen, Sulfur Dioxide |
| 17-000-0045 | LEL-I, Chlorine, Oxygen |
| 17-000-0046 | LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide |
| 17-000-0047 | LEL-I, Carbon Monoxide, Oxygen |
| 17-000-0048 | LEL-I, Oxygen, PID |
| 17-000-0049 | LEL-I, Oxygen |
| 17-000-0050 | LEL-I, PID |
| 17-000-0051 | Chlorine, Carbon Monoxide, Oxygen, LEL-P |
| 17-000-0052 | Chlorine, Oxygen, LEL-P, Cobalt Hydride |
| 17-000-0053 | Carbon Monoxide, Oxygen, LEL-P, PID |
| 17-000-0054 | Carbon Monoxide, Oxygen, LEL-P |
| 17-000-0055 | Carbon Monoxide, Oxygen |
| 17-000-0056 | Oxygen, LEL-P, Cobalt Hydride, PID |
| 17-000-0057 | Oxygen, LEL-P, PID |
| 17-000-0058 | Oxygen, LEL-P |
| 17-000-0059 | Oxygen, PID |
| 17-000-0023 | Carbon Dioxide, Oxygen |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 48 options available.
Catalog No.
17-000-0002
Supplier
Blackline Safety™
Supplier No.
G7CQACIONA
Description
- Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
- Emergency latch: Pull latch to trigger remote alert
- Silent emergency: Press and hold latch to trigger remote alert
- Low-battery: configurable threshold
- Worker check-in: Configurable timer (5 to180 min, or off ), automatic check-in when driving
Specifications
Specifications
| Components Measured | Methane, Carbon Dioxide, LEL-I, Oxygen |
| Type | Multigas Cartridge |