Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G9a/EHMT2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10040825AF488
Description
G9a/EHMT2 Polyclonal antibody specifically detects G9a/EHMT2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
G9a/EHMT2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ankyrin repeat-containing protein, BAT8, C6orf30, chromosome 6 open reading frame 30, DKFZp686H08213, EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 2HLA-B-associated transcript 8, FLJ35547, G9A histone methyltransferase, G9AEm:AF134726.3, H3-K9-HMTase 3, Histone H3-K9 methyltransferase 3, histone-lysine N-methyltransferase, H3 lysine-9 specific 3, HLA-B associated transcript 8, KMT1CNG36/G9a, Lysine N-methyltransferase 1C, NG36, Protein G9a | |
The immunogen recognized by this antibody maps to a region between residue 1160 and the C-terminus (residue 1210) of human G9A (Euchromatic Histone-Lysine N-methyltransferase 2) using the numbering given in entry NP_006700.2 (GeneID 10919). | |
0.1 mL | |
Breast Cancer | |
10919 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction