Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G9a/EHMT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10040825
Description
G9a/EHMT2 Polyclonal specifically detects G9a/EHMT2 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
G9a/EHMT2 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
ankyrin repeat-containing protein, BAT8, C6orf30, chromosome 6 open reading frame 30, DKFZp686H08213, EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 2HLA-B-associated transcript 8, FLJ35547, G9A histone methyltransferase, G9AEm:AF134726.3, H3-K9-HMTase 3, Histone H3-K9 methyltransferase 3, histone-lysine N-methyltransferase, H3 lysine-9 specific 3, HLA-B associated transcript 8, KMT1CNG36/G9a, Lysine N-methyltransferase 1C, NG36, Protein G9a | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla and Small-eared galago. | |
Store at 4C. Do not freeze. |
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
Immunohistochemistry, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:1000 -1:5000, Western Blot (Negative) | |
Q96KQ7 | |
EHMT2 | |
The immunogen recognized by this antibody maps to a region between residue 1160 and the C-terminus (residue 1210) of human G9A (Euchromatic Histone-Lysine N-methyltransferase 2) using the numbering given in entry NP_006700.2 (GeneID 10919). | |
100 μL | |
Breast Cancer | |
10919 | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction