Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G9a/EHMT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP255881
Description
G9a/EHMT2 Polyclonal specifically detects G9a/EHMT2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
G9a/EHMT2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
ankyrin repeat-containing protein, BAT8, C6orf30, chromosome 6 open reading frame 30, DKFZp686H08213, EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 2HLA-B-associated transcript 8, FLJ35547, G9A histone methyltransferase, G9AEm:AF134726.3, H3-K9-HMTase 3, Histone H3-K9 methyltransferase 3, histone-lysine N-methyltransferase, H3 lysine-9 specific 3, HLA-B associated transcript 8, KMT1CNG36/G9a, Lysine N-methyltransferase 1C, NG36, Protein G9a | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
EHMT2 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PRSEETLPKATPDSLEPAGPSSPASVTVTVGDEGADTPVGATPLIGDESENLEGDGDLRGGRILLGHATKSFPSS | |
100 μL | |
Breast Cancer | |
10919 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction