Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ G9a/EHMT2 Overexpression Lysate
SDP

Catalog No. NBL110164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBL110164 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBL110164 Supplier Novus Biologicals™ Supplier No. NBL110164
Add to Cart
Add to Cart

Applications: Western Blot, Chromatin Immunoprecipitation

The G9a/EHMT2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation.

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC2192 and based on accession number NM_25256. The protein contains a C-terminal DDK tag.
For Use With (Application) Chromatin Immunoprecipitation, Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens euchromatic histone-lysine N-methyltransferase 2 (EHMT2), transcript variant NG36/G9a-SPI, mRNA.
Host Species Human
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.