Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596756 Supplier Invitrogen™ Supplier No. PA596756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: LO2, HT-29, 293T, A-549, DU145, Mouse liver, Mouse heart, Mouse thymus, Rat liver; Cellular Location: Lysosome, Lysosome membrane.

This gene encodes acid alpha-glucosidase, which is essential for the degradation of glycogen to glucose in lysosomes. Different forms of acid alpha-glucosidase are obtained by proteolytic processing. Defects in this gene are the cause of glycogen storage disease II, also known as Pompe's disease, which is an autosomal recessive disorder with a broad clinical spectrum. Three transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAA
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gaa
Gene Accession No. P10253, P70699, Q6P7A9
Gene Alias 70 kDa lysosomal alpha-glucosidase; 76 kDa lysosomal alpha-glucosidase; acid (Pompe disease, glycogen storage disease type II); acid alpha-glucosidase; acid maltase; Aglucosidase alfa; E430018M07Rik; Gaa; glucosidase alpha, acid; glucosidase, alpha, acid; glucosidase, alpha; acid (Pompe disease, glycogen storage disease type II); LYAG; Lysosomal alpha-glucosidase
Gene Symbols Gaa
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14387, 2548, 367562
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.