Learn More
GAA Rabbit anti-Human, Polyclonal, Bioss

Description
Specifications
Specifications
| Antigen | GAA |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH |
| Gene | Gaa |
| Gene Accession No. | P10253 |
| Gene Alias | 70 kDa lysosomal alpha-glucosidase; 76 kDa lysosomal alpha-glucosidase; acid (Pompe disease, glycogen storage disease type II); acid alpha-glucosidase; Acid maltase; Aglucosidase alfa; E430018M07Rik; Gaa; glucosidase alpha, acid; glucosidase, alpha, acid; glucosidase, alpha; acid (Pompe disease, glycogen storage disease type II); LYAG; Lysosomal alpha-glucosidase |
| Gene Symbols | Gaa |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.