Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAB1 Mouse anti-Human, Clone:5F11C8, Abnova™

Catalog No. 89333114 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-114 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-114 Supplier Abnova Corporation Supplier No. MAB16664
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human GAB1.

The protein encoded by this gene is a member of the IRS1-like multisubstrate docking protein family. It is an important mediator of branching tubulogenesis and plays a central role in cellular growth response, transformation and apoptosis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen GAB1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5F11C8
Conjugate Unconjugated
Description GRB2-associated binding protein 1
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene GAB1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 661-724 of human GAB1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 2549
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.