Learn More
GAB1, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303288AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Gab1 is a 115 kDa multiple docking protein that plays an essential role in cellular growth, transformation and apoptosis. Gab1 can be phosphorylated by multiple receptor tyrosine kinase (RTKs), including: insulin receptor (IR), platelet-derived growth factor receptor beta] (PDGFRbeta]), hepatocyte growth factor/scatter factor receptor (HGFR/SFR or c-Met), and epidermal growth factor receptor (EGF), as well as in response to cell-cell adhesion. Gab1 is tyrosine phosphorylated on at least 16 sites, some of which serve as binding sites for phosphoatidylinositol 3-kinase (PI3K), Grb2, PLC gamma 1, Nck, and SHP2. Phosphorylation of Gab1 on tyrosines 627 and 659 is critical for its binding to SHP2, and for activation of the ERK/MAPK pathway in response to EGF.Specifications
GAB1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q13480 | |
GAB1 | |
Between 644 and 694 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GAB1 | |
GRB2-associated binder 1; GRB2-associated-binding protein 1; OTTHUMP00000220263; growth factor receptor bound protein 2-associated protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2549 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.